Tisk…

Methyl-6-{bis[(tert-butoxy)carbonyl]amino}pyridine-3-carboxylate

Dodavatel: Apollo Scientific

OR303641-1G OR303641-500MG OR303641-5G
APOSOR303641-1GEA 9560 CZK
APOSOR303641-1G APOSOR303641-500MG APOSOR303641-5G
Methyl-6-{bis[(tert-butoxy)carbonyl]amino}pyridine-3-carboxylate
Methyl-6-{bis[(tert-butoxy)carbonyl]amino}pyridine-3-carboxylate
Vzorec: C₁₇H₂₄N₂O₆
MW: 352,39 g/mol
Bod tání: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD18904309
CAS číslo: 666721-08-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR