tert-Butyl-4-(5-bromo-3-nitro-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate
Dodavatel: Apollo Scientific
OR303640-1G
OR303640-500MG
OR303640-5G
APOSOR303640-1GEA
9560
CZK
APOSOR303640-1G
APOSOR303640-500MG
APOSOR303640-5G
tert-Butyl-4-(5-bromo-3-nitro-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate
tert-Butyl-4-(5-bromo-3-nitro-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate
Vzorec:
C₁₄H₁₉BrN₄O₄ MW: 387,24 g/mol Bod tání: 100…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD16140319 CAS číslo: 1221792-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...