Tisk…

4-Bromophenylboronic acid pinacol ester 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromophenylboronic acid, pinacol ester, 2-(4-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Bromophenylboronic acid Pinacol ester, 1-Bromo-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

OR303515-5G OR303515-25G
APOSOR303515-5GEA 910 CZK
APOSOR303515-5G APOSOR303515-25G
4-Bromophenylboronic acid pinacol ester 97%
4-Bromophenylboronic acid pinacol ester
Vzorec: C₁₂H₁₆BBrO₂
MW: 282.97
Bod tání: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD11110553
CAS číslo: 68716-49-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR