Tisk…

4-(1-Piperidinyl)benzeneboronic acid pinacol ester

Dodavatel: Apollo Scientific

Synonyms: 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine, 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidine, 1-[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-piperidine

OR303478-1G OR303478-500MG
APOSOR303478-1GEA 2980 CZK
APOSOR303478-1G APOSOR303478-500MG
4-(1-Piperidinyl)benzeneboronic acid pinacol ester
4-(1-Piperidinyl)benzeneboronic acid pinacol ester
Vzorec: C₁₇H₂₆BNO₂
MW: 287,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368524
CAS číslo: 852227-96-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR