Tisk…

4-(Phenylethynyl)benzeneboronic acid pinacol ester

Dodavatel: Apollo Scientific

Synonyms: 4,4,5,5-Tetramethyl-2-(4-phenylethynyl-phenyl)-[1,3,2]dioxaborolane

OR303468-1G OR303468-5G
APOSOR303468-1GEA 3270 CZK
APOSOR303468-1G APOSOR303468-5G
4-(Phenylethynyl)benzeneboronic acid pinacol ester
4-(Phenylethynyl)benzeneboronic acid pinacol ester
Vzorec: C₂₀H₂₁BO₂
MW: 304,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16294498
CAS číslo: 1190376-20-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR