4-(Diethylcarbamoyl)phenylboronic acid pinacol ester 98%
Dodavatel: Apollo Scientific
Synonyms:
4-(N,N-Diethylaminocarbonyl)phenylboronic acid, pinacol ester, [4-(Diethylamine-1-carbonyl)phenyl]boronic acid pinacol ester, N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
OR303437-100G
OR303437-25G
OR303437-5G
APOSOR303437-100GEA
18350
CZK
APOSOR303437-100G
APOSOR303437-25G
APOSOR303437-5G
4-(Diethylcarbamoyl)phenylboronic acid pinacol ester 98%
4-(Diethylcarbamoyl)phenylboronic acid pinacol ester
Vzorec:
C₁₇H₂₆BNO₃ MW: 303.2000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06795673 CAS číslo: 325142-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...