Tisk…

4-Iodophenylboronic acid pinacol ester

Dodavatel: Apollo Scientific

Synonyms: 4-Iodobenzeneboronic acid pinacol ester, 2-(4-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR303432-5G
APOSOR303432-5GEA 3230 CZK
APOSOR303432-5G
4-Iodophenylboronic acid pinacol ester
4-Iodophenylboronic acid pinacol ester
Vzorec: C₁₂H₁₆BIO₂
MW: 329,97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953499
CAS číslo: 73852-88-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR