Tisk…

Methyl-1H-indazole-3-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 1H-indazole-3-carboxylate

OR303318-100G
APOSOR303318-100GEA 3150 CZK
APOSOR303318-100G
Methyl-1H-indazole-3-carboxylate
Methyl-1H-indazole-3-carboxylate
Vzorec: C₉H₈N₂O₂
MW: 176,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01138133
CAS číslo: 43120-28-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR