18-(tert-Butoxy)-18-oxooctadecanoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR303231-1G
OR303231-5G
OR303231-10G
APOSOR303231-1GEA
622
CZK
APOSOR303231-1G
APOSOR303231-5G
APOSOR303231-10G
18-(tert-Butoxy)-18-oxooctadecanoic acid
18-(tert-Butoxy)-18-oxooctadecanoic acid
Vzorec:
C₂₂H₄₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD09991735 CAS číslo: 843666-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...