(2,4-Dimethylthiazol-5-yl)acetic acid
Dodavatel: Apollo Scientific
OR303204-1G
OR303204-500MG
APOSOR303204-1GEA
9560
CZK
APOSOR303204-1G
APOSOR303204-500MG
(2,4-Dimethylthiazol-5-yl)acetic acid
(2,4-Dimethylthiazol-5-yl)acetic acid
Vzorec:
C₇H₉NO₂S MW: 171,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01681584 CAS číslo: 34272-65-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...