N-(4-Methylquinazolin-2-yl)guanidine
Dodavatel: Apollo Scientific
|
Varování
|
OR303178-1G
OR303178-500MG
APOSOR303178-1GEA
15310
CZK
APOSOR303178-1G
APOSOR303178-500MG
N-(4-Methylquinazolin-2-yl)guanidine
N-(4-Methylquinazolin-2-yl)guanidine
Vzorec:
C₁₀H₁₁N₅ MW: 201,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00023908 CAS číslo: 716-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...