Tisk…

6-Methyl-1,2,4-triazin-5-ol

Dodavatel: Apollo Scientific

OR303113-1G OR303113-500MG
APOSOR303113-1GEA 19110 CZK
APOSOR303113-1G APOSOR303113-500MG
6-Methyl-1,2,4-triazin-5-ol
6-Methyl-1,2,4-triazin-5-ol
Vzorec: C₄H₅N₃O
Storage Temperature: Ambient
MDL Number: MFCD11108305
CAS číslo: 16120-00-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR