Methyl-6-amino-2,5-dichloropyrimidine-4-carboxylate
Dodavatel: Apollo Scientific
OR303095-1G
OR303095-500MG
APOSOR303095-1GEA
19110
CZK
APOSOR303095-1G
APOSOR303095-500MG
Methyl-6-amino-2,5-dichloropyrimidine-4-carboxylate
Methyl-6-amino-2,5-dichloropyrimidine-4-carboxylate
Vzorec:
C₆H₅Cl₂N₃O₂ Bod tání: 168…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD16660543 CAS číslo: 502142-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...