Ethyl-2,6-dichloropyridine-3-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ethyl 2,6-Dichloropyridine-3-carboxylate, 2,6-Dichloropyridine-3-carboxylic acid ethyl ester, 2,6-Dichloronicotinic acid ethyl ester, Ethyl 2,6-Dichloronicotinate
OR303082-1G
OR303082-25G
OR303082-5G
APOSOR303082-1GEA
622
CZK
APOSOR303082-1G
APOSOR303082-25G
APOSOR303082-5G
Ethyl-2,6-dichloropyridine-3-carboxylate
Ethyl-2,6-dichloropyridine-3-carboxylate
Vzorec:
C₈H₇Cl₂NO₂ MW: 220,06 g/mol Bod tání: 50…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD09757510 CAS číslo: 58584-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...