(2,4-Dioxo-1,2,3,4-tetrahydropyrimidin-5-ylamino)fumaric acid dimethyl ester
Dodavatel: Apollo Scientific
OR303078-1G
OR303078-500MG
APOSOR303078-1GEA
5130
CZK
APOSOR303078-1G
APOSOR303078-500MG
(2,4-Dioxo-1,2,3,4-tetrahydropyrimidin-5-ylamino)fumaric acid dimethyl ester
(2,4-Dioxo-1,2,3,4-tetrahydropyrimidin-5-ylamino)fumaric acid dimethyl ester
Vzorec:
C₁₀H₁₁N₃O₆ MW: 269,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22580727 CAS číslo: 60458-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...