Tisk…

4-Bromo-N-(1,3-thiazol-2-yl)benzene-1-sulphonamide

Dodavatel: Apollo Scientific

OR303065-1G OR303065-500MG
APOSOR303065-1GEA 9560 CZK
APOSOR303065-1G APOSOR303065-500MG
4-Bromo-N-(1,3-thiazol-2-yl)benzene-1-sulphonamide
4-Bromo-N-(1,3-thiazol-2-yl)benzene-1-sulphonamide
Vzorec: C₉H₇BrN₂O₂S₂
MW: 319,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00592646
CAS číslo: 331972-47-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR