5-Bromo-[1,3]thiazolo[5,4-b]pyridin-2-amine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Bromothiazolo[5,4-b]pyridin-2-amine
OR303064-1G
OR303064-250MG
OR303064-5G
APOSOR303064-1GEA
3150
CZK
APOSOR303064-1G
APOSOR303064-250MG
APOSOR303064-5G
5-Bromo-[1,3]thiazolo[5,4-b]pyridin-2-amine
5-Bromo-[1,3]thiazolo[5,4-b]pyridin-2-amine
Vzorec:
C₆H₄BrN₃S MW: 230,09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11846492 CAS číslo: 934266-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...