Tisk…

4-Bromo-N-(1,3,4-thiadiazol-2-yl)benzene-1-sulphonamide

Dodavatel: Apollo Scientific

OR303063-1G OR303063-500MG
APOSOR303063-1GEA 15310 CZK
APOSOR303063-1G APOSOR303063-500MG
4-Bromo-N-(1,3,4-thiadiazol-2-yl)benzene-1-sulphonamide
4-Bromo-N-(1,3,4-thiadiazol-2-yl)benzene-1-sulphonamide
Vzorec: C₈H₆BrN₃O₂S₂
MW: 320,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02861948
CAS číslo: 349614-28-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR