Tisk…

N-Phenylmaleimide 98%

Dodavatel: Apollo Scientific
Nebezpečí

OR30270-500G
APOSOR30270-500GEA 0 CZK
APOSOR30270-500G
N-Phenylmaleimide 98%
N-Phenylmaleimide
Vzorec: C₁₀H₇NO₂
MW: 173,17 g/mol
Bod varu: 162…163 °C/12mmHg
Bod tání: 85…87 °C
Hustota: 1,18 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005502
CAS číslo: 941-69-5
EINECS: 213-382-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR