Tisk…

2,4-Dichloro-6-methylbenzenesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,4-Dichloro-6-methylbenzene-1-sulphonyl chloride

OR30237-1G OR30237-500MG OR30237-5G
APOSOR30237-1GEA 3230 CZK
APOSOR30237-1G APOSOR30237-500MG APOSOR30237-5G
2,4-Dichloro-6-methylbenzenesulphonyl chloride
2,4-Dichloro-6-methylbenzenesulphonyl chloride
Vzorec: C₇H₅Cl₃O₂S
MW: 259,54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00204179
CAS číslo: 175278-26-9
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR