Tisk…

2-Amino-4-iodopyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Iodopyridin-2-amine

OR302288-5G
APOSOR302288-5GEA 2150 CZK
APOSOR302288-5G
2-Amino-4-iodopyridine
2-Amino-4-iodopyridine
Vzorec: C₅H₅IN₂
MW: 220,01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09038163
CAS číslo: 552331-00-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR