Triphenylacetic acid 97%
Dodavatel: Apollo Scientific
OR302273-5G
OR302273-25G
APOSOR302273-5GEA
1660
CZK
APOSOR302273-5G
APOSOR302273-25G
Triphenylacetic acid 97%
Triphenylacetic acid
Vzorec:
C₂₀H₁₆O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00004185 CAS číslo: 595-91-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...