6-Methoxy-2-methyl-3-nitropyridine
Dodavatel: Apollo Scientific
OR302187-25G
APOSOR302187-25GEA
2690
CZK
APOSOR302187-25G
6-Methoxy-2-methyl-3-nitropyridine
6-Methoxy-2-methyl-3-nitropyridine
Vzorec:
C₇H₈N₂O₃ MW: 168,1500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03095076 CAS číslo: 5467-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...