4-Bromo-1-(2-tetrahydropyranyl)-1H-pyrazole
Dodavatel: Apollo Scientific
Synonyms:
4-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole, 4-Bromo-1-(tetrahydro-2H-pyran-2-yl-1H)-pyrazole
OR302129-5G
OR302129-25G
APOSOR302129-5GEA
744
CZK
APOSOR302129-5G
APOSOR302129-25G
4-Bromo-1-(2-tetrahydropyranyl)-1H-pyrazole
4-Bromo-1-(2-tetrahydropyranyl)-1H-pyrazole
Vzorec:
C₈H₁₁BrN₂O MW: 231,0900 g/mol Bod varu: 115 °C Hustota: 1,493 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD12026052 CAS číslo: 82099-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...