Imidazo[1,5-a]pyridine-8-carboxylic acid 98%
Dodavatel: Apollo Scientific
OR301331-100MG
OR301331-1G
OR301331-250MG
APOSOR301331-100MGEA
8500
CZK
APOSOR301331-100MG
APOSOR301331-1G
APOSOR301331-250MG
Imidazo[1,5-a]pyridine-8-carboxylic acid 98%
Imidazo[1,5-a]pyridine-8-carboxylic acid
Vzorec:
C₈H₆N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD11870762 CAS číslo: 697739-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...