(S)-(+)-2-Amino-1-phenylethanol 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR3013-1G
OR3013-5G
APOSOR3013-1GEA
1080
CZK
APOSOR3013-1G
APOSOR3013-5G
(S)-(+)-2-Amino-1-phenylethanol 98%
(S)-(+)-2-Amino-1-phenylethanol
Vzorec:
C₈H₁₁NO MW: 137,182 g/mol Bod varu: 160 °C/17mmHg Bod tání: 57…63 °C Storage Temperature: Freezer |
MDL Number:
MFCD00239405 CAS číslo: 56613-81-1 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...