Tisk…

4-Methyl-5-nitro-1H-indole

Dodavatel: Apollo Scientific

OR301286-1G OR301286-250MG
APOSOR301286-1GEA 2440 CZK
APOSOR301286-1G APOSOR301286-250MG
4-Methyl-5-nitro-1H-indole
4-Methyl-5-nitro-1H-indole
Vzorec: C₉H₈N₂O₂
MW: 176.1720 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11035915
CAS číslo: 165250-69-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR