Tisk…

Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate 95%

Dodavatel: Apollo Scientific

Synonyms: Methyl 1H-pyrrolo[2,3-b]pyridine-5-carboxylate

OR301280-5G
APOSOR301280-5GEA 3940 CZK
APOSOR301280-5G
Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate 95%
Methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate
Vzorec: C₉H₈N₂O₂
MW: 176,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08457785
CAS číslo: 849067-96-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR