Tisk…

4-Bromo-3-iodo-7-azaindole 95+%

Dodavatel: Apollo Scientific

OR301273-1G OR301273-250MG OR301273-5G
APOSOR301273-1GEA 3740 CZK
APOSOR301273-1G APOSOR301273-250MG APOSOR301273-5G
4-Bromo-3-iodo-7-azaindole 95+%
4-Bromo-3-iodo-7-azaindole
Vzorec: C₇H₄BrIN₂
Storage Temperature: Ambient
MDL Number: MFCD09880123
CAS číslo: 1000340-34-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR