Tisk…

4-Nitro-7-azaindole 98%

Dodavatel: Apollo Scientific

OR301264-1G OR301264-5G
APOSOR301264-1GEA 2240 CZK
APOSOR301264-1G APOSOR301264-5G
4-Nitro-7-azaindole 98%
4-Nitro-7-azaindole
Vzorec: C₇H₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD07778365
CAS číslo: 83683-82-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR