Tisk…

N-Boc-N-methylethylenediamine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: N-Boc-(2-Aminoethyl)-N-methylamine, N-Methylethane-1,2-diamine, N-BOC protected

OR301251-1G OR301251-5G
APOSOR301251-1GEA 622 CZK
APOSOR301251-1G APOSOR301251-5G
N-Boc-N-methylethylenediamine
N-Boc-N-methylethylenediamine
Vzorec: C₈H₁₈N₂O₂
MW: 174,24 g/mol
Hustota: 0,975 g/cm³ (25 °C)
Bod vzplanutí: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD01317789
CAS číslo: 121492-06-6
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR