Tisk…

2-Amino-3-methylpyrazine 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Methylpyrazin-2-amine

OR301244-5G
APOSOR301244-5GEA 2690 CZK
APOSOR301244-5G
2-Amino-3-methylpyrazine 97%
2-Amino-3-methylpyrazine
Vzorec: C₅H₇N₃
MW: 109,132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08705770
CAS číslo: 19838-08-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR