Tisk…

(2-Chloropyridin-4-yl)methanamine hydrochloride 95+%

Dodavatel: Apollo Scientific
Varování

OR301242-1G OR301242-100MG OR301242-250MG
APOSOR301242-1GEA 4570 CZK
APOSOR301242-1G APOSOR301242-100MG APOSOR301242-250MG
(2-Chloropyridin-4-yl)methanamine hydrochloride 95+%
(2-Chloropyridin-4-yl)methanamine hydrochloride
Vzorec: C₆H₈Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD21362508
CAS číslo: 916210-98-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR