2-Amino-4-bromopyrimidine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Bromopyrimidin-2-amine
OR301239-1G
OR301239-5G
APOSOR301239-1GEA
2440
CZK
APOSOR301239-1G
APOSOR301239-5G
2-Amino-4-bromopyrimidine
2-Amino-4-bromopyrimidine
Vzorec:
C₄H₄BrN₃ MW: 174,00 g/mol Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD09999168 CAS číslo: 343926-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...