(2-Methoxypyridin-4-yl)methanol 98%
Dodavatel: Apollo Scientific
OR301205-1G
OR301205-5G
APOSOR301205-1GEA
817
CZK
APOSOR301205-1G
APOSOR301205-5G
(2-Methoxypyridin-4-yl)methanol 98%
(2-Methoxypyridin-4-yl)methanol
Vzorec:
C₇H₉NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00859298 CAS číslo: 123148-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...