Tisk…

1H-Pyrazole-3-boronic acid pinacol ester 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 2-(1H-Pyrazol-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1H-Pyrazole-3-boronic acid Pinacol ester

OR301189-100G OR301189-25G
APOSOR301189-100GEA 7110 CZK
APOSOR301189-100G APOSOR301189-25G
1H-Pyrazole-3-boronic acid pinacol ester 97%
1H-Pyrazole-3-boronic acid pinacol ester
Vzorec: C₉H₁₅BN₂O₂
MW: 194,04 g/mol
Bod tání: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD07368044
CAS číslo: 844501-71-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR