7-Bromo-2-naphthol
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
7-Bromonaphthalen-2-ol, 7-Bromo-2-hydroxynaphthalene, 2-Bromo-7-hydroxynaphthalene
OR301112-1G
OR301112-10G
APOSOR301112-1GEA
622
CZK
APOSOR301112-1G
APOSOR301112-10G
7-Bromo-2-naphthol
7-Bromo-2-naphthol
Vzorec:
C₁₀H₇BrO MW: 223,07 g/mol Bod tání: 133…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00277644 CAS číslo: 116230-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...