Tisk…

6-Bromo-3-nitro-4-quinolinol

Dodavatel: Apollo Scientific

Synonyms: 6-Bromo-3-nitroquinolin-4-ol

OR301107-5G
APOSOR301107-5GEA 4800 CZK
APOSOR301107-5G
6-Bromo-3-nitro-4-quinolinol
6-Bromo-3-nitro-4-quinolinol
Vzorec: C₉H₅BrN₂O₃
MW: 269,0516 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800413
CAS číslo: 853908-50-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR