(R)-2-Amino-1-phenylethanol
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
(R)-(-)-2-Amino-1-phenylethanol, (R)-(-)-alpha-(Aminomethyl)benzyl alcohol
OR3011-1G
APOSOR3011-1GEA
744
CZK
APOSOR3011-1G
(R)-2-Amino-1-phenylethanol
(R)-2-Amino-1-phenylethanol
Vzorec:
C₈H₁₁NO MW: 137,18 g/mol Bod varu: 160 °C/17mmHg Bod tání: 57…65 °C Storage Temperature: Freezer |
MDL Number:
MFCD00239406 CAS číslo: 2549-14-6 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...