4-Amino-6-chloropyrimidine-5-carbonitrile
Dodavatel: Apollo Scientific
|
Varování
|
OR301091-25G
OR301091-5G
APOSOR301091-25GEA
7870
CZK
APOSOR301091-25G
APOSOR301091-5G
4-Amino-6-chloropyrimidine-5-carbonitrile
4-Amino-6-chloropyrimidine-5-carbonitrile
Vzorec:
C₅H₃ClN₄ MW: 154,5600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07761812 CAS číslo: 60025-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...