Tisk…

tert-Butyl (3-oxocyclohexyl)carbamate 97%

Dodavatel: Apollo Scientific
Varování

OR301088-1G OR301088-5G
APOSOR301088-1GEA 622 CZK
APOSOR301088-1G APOSOR301088-5G
tert-Butyl (3-oxocyclohexyl)carbamate 97%
tert-Butyl (3-oxocyclohexyl)carbamate
Vzorec: C₁₁H₁₉NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD03790905
CAS číslo: 885280-38-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR