2-Methylmorpholine
Dodavatel: Apollo Scientific
|
Varování
|
OR301073-1G
OR301073-250MG
APOSOR301073-1GEA
2820
CZK
APOSOR301073-1G
APOSOR301073-250MG
2-Methylmorpholine
2-Methylmorpholine
Vzorec:
C₅H₁₁NO MW: 101,1469 g/mol Bod varu: 136 °C Storage Temperature: Ambient |
MDL Number:
MFCD08444368 CAS číslo: 27550-90-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...