Tisk…

2-Bromo-p-anisaldehyde 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromo-4-methoxybenzaldehyde

OR301068-25G OR301068-100G OR301068-5G
APOSOR301068-25GEA 2400 CZK
APOSOR301068-25G APOSOR301068-100G APOSOR301068-5G
2-Bromo-p-anisaldehyde 98%
2-Bromo-p-anisaldehyde
Vzorec: C₈H₇BrO₂
MW: 215,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09788636
CAS číslo: 43192-31-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR