3-Aminocyclohexanol
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR301031-5G
OR301031-25G
APOSOR301031-5GEA
2650
CZK
APOSOR301031-5G
APOSOR301031-25G
3-Aminocyclohexanol
3-Aminocyclohexanol
Vzorec:
C6H13NO MW: 115,1700 g/mol Bod varu: 118…122 °C/13mmHg Bod tání: 64…69 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03790923 CAS číslo: 6850-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...