Tisk…

3-Bromo-2-chloro-4-methylpyridine 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Bromo-2-chloro-4-picoline

OR301029-5G OR301029-100G OR301029-25G
APOSOR301029-5GEA 3310 CZK
APOSOR301029-5G APOSOR301029-100G APOSOR301029-25G
3-Bromo-2-chloro-4-methylpyridine 97%
3-Bromo-2-chloro-4-methylpyridine
Vzorec: C₆H₅BrClN
MW: 206.47
Storage Temperature: Ambient
MDL Number: MFCD04112511
CAS číslo: 55404-31-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR