Tisk…

2-Amino-5-pyridineboronic acid pinacol ester 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Aminopyridine-3-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine, 2-Aminopyridine-5-boronic acid pinacol ester

OR3010-5G OR3010-25G
APOSOR3010-5GEA 2980 CZK
APOSOR3010-5G APOSOR3010-25G
2-Amino-5-pyridineboronic acid pinacol ester 95%
2-Amino-5-pyridineboronic acid pinacol ester
Vzorec: C₁₁H₁₇BN₂O₂
MW: 220.08
Bod tání: 131…135 °C
Storage Temperature: Ambient
MDL Number: MFCD05663837
CAS číslo: 827614-64-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR