3-Methoxybenzamidoxime
Dodavatel: Apollo Scientific
OR300766-1G
OR300766-250MG
OR300766-5G
APOSOR300766-1GEA
4470
CZK
APOSOR300766-1G
APOSOR300766-250MG
APOSOR300766-5G
3-Methoxybenzamidoxime
3-Methoxybenzamidoxime
Vzorec:
C₈H₁₀N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD07161438 CAS číslo: 73647-50-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...