Tisk…

3-(2-Methylthiazol-4-yl)benzenesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR300764-1G OR300764-5G OR300764-10G
APOSOR300764-1GEA 0 CZK
APOSOR300764-1G APOSOR300764-5G APOSOR300764-10G
3-(2-Methylthiazol-4-yl)benzenesulphonyl chloride
3-(2-Methylthiazol-4-yl)benzenesulphonyl chloride
Vzorec: C₁₀H₈ClNO₂S₂
Bod tání: 116…118 °C
Storage Temperature: Ambient
MDL Number: MFCD07772796
CAS číslo: 66047-75-4
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR