Tisk…

Ethyl-3-amino-4,6-dimethylthieno[2,3-b]pyridine-2-carboxylate

Dodavatel: Apollo Scientific

OR300731-1G OR300731-50MG OR300731-250MG OR300731-5G
APOSOR300731-1GEA 2820 CZK
APOSOR300731-1G APOSOR300731-50MG APOSOR300731-250MG APOSOR300731-5G
Ethyl-3-amino-4,6-dimethylthieno[2,3-b]pyridine-2-carboxylate
Ethyl-3-amino-4,6-dimethylthieno[2,3-b]pyridine-2-carboxylate
Vzorec: C₁₂H₁₄N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD00124672
CAS číslo: 52505-56-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR