Tisk…

4-Methylnaphthalene-1-sulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR30073-1G OR30073-5G
APOSOR30073-1GEA 2400 CZK
APOSOR30073-1G APOSOR30073-5G
4-Methylnaphthalene-1-sulphonyl chloride
4-Methylnaphthalene-1-sulphonyl chloride
Vzorec: C₁₁H₉ClO₂S
MW: 240,7060 g/mol
Bod tání: 81…82 °C
Storage Temperature: Ambient
MDL Number: MFCD02677693
CAS číslo: 10447-11-7
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR